Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTCGCCAGGCCCTTACCTGAGC[C/T]GATGCGGGCCCGAGACATGGTGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107310 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC9A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC9A1 - solute carrier family 9 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003047.4 | 1978 | Missense Mutation | AGC,GGC | S,G 702 | NP_003038.2 | |
XM_011542021.2 | 1978 | Missense Mutation | AGC,GGC | S,G 592 | XP_011540323.1 |