Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAATGCCATCGCCATTGTGCAGCT[A/G]CAGGAACTCTCTTTGAGGCTGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120420 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSF1 - colony stimulating factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000757.5 | 749 | Silent Mutation | CTA,CTG | L,L 112 | NP_000748.3 | |
NM_172210.2 | 749 | Silent Mutation | CTA,CTG | L,L 112 | NP_757349.1 | |
NM_172211.3 | 749 | Silent Mutation | CTA,CTG | L,L 112 | NP_757350.1 | |
NM_172212.2 | 749 | Silent Mutation | CTA,CTG | L,L 112 | NP_757351.1 | |
XM_017000369.1 | 749 | Silent Mutation | CTA,CTG | L,L 71 | XP_016855858.1 |