Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGTCCTCATACTTGTACTTGCCG[A/G]TCAGCAGGCCCCCTGAGGGAAAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608477 | ||||||||||||||||||||
Literature Links: |
AKR7A3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKR7A3 - aldo-keto reductase family 7 member A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012067.2 | 428 | Missense Mutation | ACC,ATC | T,I 206 | NP_036199.2 | |
XM_011541046.2 | 428 | Missense Mutation | ACC,ATC | T,I 206 | XP_011539348.1 | |
XM_017000714.1 | 428 | Missense Mutation | ACC,ATC | T,I 135 | XP_016856203.1 |
LOC100506730 - uncharacterized LOC100506730 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |