Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGTTGGCATCTGGTCTTGACAC[C/T]GCAGAGAAGGCCTTGAAAGTTGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC30 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC30 - coiled-coil domain containing 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080850.2 | 1129 | Silent Mutation | ACC,ACT | T,T 29 | NP_001074319.1 | |
XM_011542067.2 | 1129 | Silent Mutation | ACC,ACT | T,T 29 | XP_011540369.1 | |
XM_011542068.2 | 1129 | Silent Mutation | ACC,ACT | T,T 29 | XP_011540370.1 | |
XM_011542069.2 | 1129 | Silent Mutation | ACC,ACT | T,T 29 | XP_011540371.1 | |
XM_011542070.2 | 1129 | Intron | XP_011540372.1 | |||
XM_011542071.2 | 1129 | Silent Mutation | ACC,ACT | T,T 29 | XP_011540373.1 | |
XM_017002225.1 | 1129 | Silent Mutation | ACC,ACT | T,T 29 | XP_016857714.1 | |
XM_017002226.1 | 1129 | Silent Mutation | ACC,ACT | T,T 29 | XP_016857715.1 |