Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATAAGCGGTTCAACTTGGGATA[C/T]GCAAGGAGTCTCTGCAGAGGTACGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612178 | ||||||||||||||||||||
Literature Links: |
FAM102B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM102B - family with sequence similarity 102 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HENMT1 - HEN1 methyltransferase homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102592.1 | 1465 | Silent Mutation | GCA,GCG | A,A 348 | NP_001096062.1 | |
NM_144584.2 | 1465 | Silent Mutation | GCA,GCG | A,A 348 | NP_653185.2 | |
XM_005270411.1 | 1465 | Silent Mutation | GCA,GCG | A,A 356 | XP_005270468.1 | |
XM_017000179.1 | 1465 | Silent Mutation | GCA,GCG | A,A 192 | XP_016855668.1 |