Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCCAGTGGGGTCCCGGAGGTGG[A/G]GCTACGGGATGTTGCTTCGGAATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 139370 MIM: 606916 | ||||||||||||||||||||
Literature Links: |
GNAI3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GNAI3 - G protein subunit alpha i3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPR61 - G protein-coupled receptor 61 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031936.4 | 3120 | Missense Mutation | GAG,GGG | E,G 35 | NP_114142.3 | |
XM_017002438.1 | 3120 | Missense Mutation | GAG,GGG | E,G 35 | XP_016857927.1 |