Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATCTATCAGATCATCCAGCTCCT[C/G]GGAAGGTGTCAACTGCTGATGTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF281 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF281 - zinc finger protein 281 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281293.1 | 3799 | Missense Mutation | CAG,GAG | Q,E 763 | NP_001268222.1 | |
NM_001281294.1 | 3799 | Missense Mutation | CAG,GAG | Q,E 727 | NP_001268223.1 | |
NM_012482.4 | 3799 | Missense Mutation | CAG,GAG | Q,E 763 | NP_036614.1 | |
XM_017000887.1 | 3799 | Missense Mutation | CAG,GAG | Q,E 763 | XP_016856376.1 | |
XM_017000888.1 | 3799 | Missense Mutation | CAG,GAG | Q,E 763 | XP_016856377.1 |