Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTTAGACGTCGGGCATTGTCCA[A/G]GCCTCCCTTGGGATCATAGGCACGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 138290 | ||||||||||||||||||||
Literature Links: |
GLUL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLUL - glutamate-ammonia ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033044.3 | 1484 | Silent Mutation | CTG,TTG | L,L 294 | NP_001028216.1 | |
NM_001033056.3 | 1484 | Silent Mutation | CTG,TTG | L,L 294 | NP_001028228.1 | |
NM_002065.6 | 1484 | Silent Mutation | CTG,TTG | L,L 294 | NP_002056.2 | |
XM_006711278.1 | 1484 | Silent Mutation | CTG,TTG | L,L 294 | XP_006711341.1 |