Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCATGGTGCCTCCACAGGCGCAG[C/T]GAGCGGTTTTCTGGCCCCCGCTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609318 MIM: 604718 | ||||||||||||||||||||
Literature Links: |
TRIM45 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIM45 - tripartite motif containing 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145635.1 | 2156 | Missense Mutation | NP_001139107.1 | |||
NM_025188.3 | 2156 | Missense Mutation | NP_079464.2 | |||
XM_011542199.2 | 2156 | Missense Mutation | XP_011540501.1 |
TTF2 - transcription termination factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |