Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGGCAAGCAGCTGTGCTCACAGA[G/T]GCTCACTGCCAAAGTTGCTTTTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602202 MIM: 605037 | ||||||||||||||||||||
Literature Links: |
DDOST PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDOST - dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIF17 - kinesin family member 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122819.2 | 3391 | Missense Mutation | CAT,CCT | H,P 1027 | NP_001116291.1 | |
NM_001287212.1 | 3391 | Missense Mutation | CAT,CCT | H,P 928 | NP_001274141.1 | |
NM_020816.3 | 3391 | Missense Mutation | CAT,CCT | H,P 1028 | NP_065867.2 | |
XM_005245950.3 | 3391 | Intron | XP_005246007.1 | |||
XM_005245951.3 | 3391 | Intron | XP_005246008.1 | |||
XM_011541837.1 | 3391 | Intron | XP_011540139.1 | |||
XM_011541838.1 | 3391 | Intron | XP_011540140.1 | |||
XM_011541839.2 | 3391 | Intron | XP_011540141.1 | |||
XM_011541840.2 | 3391 | Intron | XP_011540142.1 | |||
XM_011541841.2 | 3391 | Intron | XP_011540143.1 | |||
XM_011541842.1 | 3391 | Intron | XP_011540144.1 | |||
XM_011541843.1 | 3391 | Intron | XP_011540145.1 | |||
XM_011541844.2 | 3391 | Intron | XP_011540146.1 | |||
XM_011541845.2 | 3391 | Intron | XP_011540147.1 | |||
XM_011541846.2 | 3391 | Intron | XP_011540148.1 | |||
XM_011541847.2 | 3391 | Intron | XP_011540149.1 |