Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAGGAGCAAGTGCTCATCAACC[A/G]CCAGGAAATCACGAACAAAGCGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609121 MIM: 603272 | ||||||||||||||||||||
Literature Links: |
CATSPER4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CATSPER4 - cation channel sperm associated 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198137.1 | 191 | Missense Mutation | CAC,CGC | H,R 64 | NP_937770.1 | |
XM_011541432.2 | 191 | Missense Mutation | CAC,CGC | H,R 64 | XP_011539734.1 | |
XM_011541433.2 | 191 | Missense Mutation | CAC,CGC | H,R 64 | XP_011539735.1 |
CNKSR1 - connector enhancer of kinase suppressor of Ras 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297647.1 | 191 | Intron | NP_001284576.1 | |||
NM_001297648.1 | 191 | Intron | NP_001284577.1 | |||
NM_006314.2 | 191 | Intron | NP_006305.2 | |||
XM_011540484.2 | 191 | Intron | XP_011538786.1 | |||
XM_011540485.2 | 191 | Intron | XP_011538787.1 |