Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGTGAACCACAACAAGTTTCTG[G/T]CTACCACAGACTACTTGCTACATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIEXF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DIEXF - digestive organ expansion factor homolog (zebrafish) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014388.6 | 309 | Missense Mutation | GGC,GTC | G,V 71 | NP_055203.4 | |
XM_006711275.3 | 309 | Missense Mutation | GGC,GTC | G,V 71 | XP_006711338.1 | |
XM_011509418.2 | 309 | UTR 5 | XP_011507720.1 |