Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGCGCGAGCAAGATGGCCACCAC[A/C]AAGCGCGTCTTGTACGTGGGTGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602435 | ||||||||||||||||||||
Literature Links: |
PPIE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPIE - peptidylprolyl isomerase E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195007.1 | 68 | Intron | NP_001181936.1 | |||
NM_001319293.1 | 68 | Intron | NP_001306222.1 | |||
NM_006112.3 | 68 | Silent Mutation | ACA,ACC | T,T 4 | NP_006103.1 | |
NM_203456.2 | 68 | Intron | NP_982281.1 | |||
XM_006710289.2 | 68 | Intron | XP_006710352.1 | |||
XM_006710290.3 | 68 | Silent Mutation | ACA,ACC | T,T 4 | XP_006710353.1 | |
XM_011540501.1 | 68 | Intron | XP_011538803.1 | |||
XM_017000051.1 | 68 | Intron | XP_016855540.1 | |||
XM_017000052.1 | 68 | UTR 5 | XP_016855541.1 |