Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGCATAAAACCATTCTTCAGAC[A/G]ACTGAAGATGGTCCCCTTCTGTAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600722 | ||||||||||||||||||||
Literature Links: |
CAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAP1 - adenylate cyclase associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPT1 - palmitoyl-protein thioesterase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000310.3 | 1107 | Missense Mutation | TCG,TTG | S,L 292 | NP_000301.1 | |
NM_001142604.1 | 1107 | Missense Mutation | TCG,TTG | S,L 189 | NP_001136076.1 | |
XM_005271008.1 | 1107 | Missense Mutation | TCG,TTG | S,L 268 | XP_005271065.1 |