Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCATCTCCTTGGGCAGGCCCTGG[A/G]CGAGGGATGGGCACTGTTTCTCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609109 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO42 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO42 - F-box protein 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018994.2 | 2063 | Silent Mutation | CGC,CGT | R,R 603 | NP_061867.1 | |
XM_006710698.3 | 2063 | Silent Mutation | CGC,CGT | R,R 603 | XP_006710761.1 | |
XM_017001502.1 | 2063 | Silent Mutation | CGC,CGT | R,R 372 | XP_016856991.1 | |
XM_017001503.1 | 2063 | Silent Mutation | CGC,CGT | R,R 372 | XP_016856992.1 |