Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGTCTCTCACGTTCGGTGCTCA[C/T]GTTGGGAAGAGGACGCTGCTCCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C1orf198 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf198 - chromosome 1 open reading frame 198 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136494.1 | 874 | Missense Mutation | ATG,GTG | M,V 220 | NP_001129966.1 | |
NM_001136495.1 | 874 | Missense Mutation | ATG,GTG | M,V 128 | NP_001129967.1 | |
NM_032800.2 | 874 | Missense Mutation | ATG,GTG | M,V 258 | NP_116189.1 | |
XM_017002599.1 | 874 | Missense Mutation | ATG,GTG | M,V 128 | XP_016858088.1 |