Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATGCCCAAATTTATTCGTTTTT[C/T]ACATGATAGCGTGAGAATTAAAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
AADACL4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AADACL4 - arylacetamide deacetylase like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013630.1 | 224 | Missense Mutation | TCA,TTA | S,L 75 | NP_001013652.1 | |
XM_017001152.1 | 224 | Missense Mutation | TCA,TTA | S,L 8 | XP_016856641.1 | |
XM_017001153.1 | 224 | Intron | XP_016856642.1 |