Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGTCTGGGAGAACTTGCCATTG[A/G]GTTTGAGGATGCCTTTGCGATGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608131 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUAK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUAK2 - NUAK family kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030952.2 | 931 | Missense Mutation | CCC,CTC | P,L 548 | NP_112214.2 | |
XM_005245515.4 | 931 | Missense Mutation | CCC,CTC | P,L 257 | XP_005245572.1 |