Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACACCAACCAGCATCATGTCCAT[A/G]ACACTGGGGTACTGGGACATCCGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 138333 | ||||||||||||||||||||
Literature Links: |
GSTM4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSTM4 - glutathione S-transferase mu 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000850.4 | 323 | Missense Mutation | ATA,ATG | I,M 3 | NP_000841.1 | |
NM_147148.2 | 323 | Missense Mutation | ATA,ATG | I,M 3 | NP_671489.1 | |
XM_017001085.1 | 323 | Missense Mutation | ATA,ATG | I,M 3 | XP_016856574.1 | |
XM_017001086.1 | 323 | UTR 5 | XP_016856575.1 | |||
XM_017001087.1 | 323 | UTR 5 | XP_016856576.1 | |||
XM_017001088.1 | 323 | UTR 5 | XP_016856577.1 |