Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGTGGGGGGCCCCTGGGGTGAG[A/C]GACGGTCCAATGCCTTCAATAAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602344 | ||||||||||||||||||||
Literature Links: |
PAFAH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAFAH2 - platelet activating factor acetylhydrolase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000437.3 | 1073 | Silent Mutation | NP_000428.2 | |||
XM_006710669.3 | 1073 | Silent Mutation | XP_006710732.1 | |||
XM_006710670.3 | 1073 | Silent Mutation | XP_006710733.1 | |||
XM_011541528.2 | 1073 | Silent Mutation | XP_011539830.1 | |||
XM_011541529.2 | 1073 | Silent Mutation | XP_011539831.1 | |||
XM_017001390.1 | 1073 | Silent Mutation | XP_016856879.1 | |||
XM_017001391.1 | 1073 | Silent Mutation | XP_016856880.1 | |||
XM_017001392.1 | 1073 | Silent Mutation | XP_016856881.1 |