Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGACCAGAAGCTCTCTGAGTTAGA[C/T]GACCGTGCAGACGCACTGCAGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611501 MIM: 603427 MIM: 603657 | ||||||||||||||||||||
Literature Links: |
CAMTA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAMTA1 - calmodulin binding transcription activator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PER3 - period circadian clock 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VAMP3 - vesicle associated membrane protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004781.3 | 250 | Silent Mutation | GAC,GAT | D,D 47 | NP_004772.1 | |
XM_017002841.1 | 250 | Silent Mutation | GAC,GAT | D,D 47 | XP_016858330.1 |