Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGAATCCATATGCTGTGGCAGGG[C/T]GTCCAAGAATGCCAACTGGTTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601836 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIFAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIFAP3 - kinesin associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204514.1 | 2753 | Missense Mutation | CAC,CGC | H,R 695 | NP_001191443.1 | |
NM_001204516.1 | 2753 | Missense Mutation | CAC,CGC | H,R 729 | NP_001191445.1 | |
NM_001204517.1 | 2753 | Missense Mutation | CAC,CGC | H,R 733 | NP_001191446.1 | |
NM_014970.3 | 2753 | Missense Mutation | CAC,CGC | H,R 773 | NP_055785.2 | |
XM_005244970.1 | 2753 | Silent Mutation | ACA,ACG | T,T 785 | XP_005245027.1 | |
XM_011509307.2 | 2753 | UTR 3 | XP_011507609.2 |