Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCACGTGGTCCCGGTCGTAGCCCC[A/G]GCCTCCTGGGGGTGGGATGGGGATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 114280 MIM: 609151 | ||||||||||||||||||||
Literature Links: |
AIM1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AIM1L - absent in melanoma 1-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039775.3 | 5039 | Missense Mutation | CGG,TGG | R,W 1635 | NP_001034864.2 | |
XM_005245918.2 | 5039 | Missense Mutation | CGG,TGG | R,W 1635 | XP_005245975.1 | |
XM_011541672.1 | 5039 | Missense Mutation | CGG,TGG | R,W 1623 | XP_011539974.1 | |
XM_011541673.2 | 5039 | Missense Mutation | CGG,TGG | R,W 1692 | XP_011539975.1 |
CD52 - CD52 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBXN11 - UBX domain protein 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |