Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGGAGCCACCATGTGGCGATGT[C/T]CACTGGGGCTACTGCTGTTGCTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616064 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TINAGL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TINAGL1 - tubulointerstitial nephritis antigen like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204414.1 | 139 | Missense Mutation | CCA,TCA | P,S 5 | NP_001191343.1 | |
NM_001204415.1 | 139 | Intron | NP_001191344.1 | |||
NM_022164.2 | 139 | Missense Mutation | CCA,TCA | P,S 5 | NP_071447.1 | |
XM_005271106.2 | 139 | Missense Mutation | CCA,TCA | P,S 5 | XP_005271163.1 | |
XM_005271107.2 | 139 | Intron | XP_005271164.1 | |||
XM_011541946.1 | 139 | Intron | XP_011540248.1 |