Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTATAGCAGCAGAAACTGACTT[A/G]TAAGTAGAGAGCTCTTCAGCAAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602923 MIM: 611750 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBBP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBBP4 - RB binding protein 4, chromatin remodeling factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135255.1 | 5892 | UTR 3 | NP_001128727.1 | |||
NM_001135256.1 | 5892 | UTR 3 | NP_001128728.1 | |||
NM_005610.2 | 5892 | UTR 3 | NP_005601.1 |
SYNC - syncoilin, intermediate filament protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161708.1 | 5892 | Silent Mutation | TAC,TAT | Y,Y 452 | NP_001155180.1 | |
NM_030786.2 | 5892 | Silent Mutation | TAC,TAT | Y,Y 452 | NP_110413.2 |