Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGTAGTGCCTTCTCTTCTCGCG[C/T]CTTCTGAGAGCTACCAGCCGGGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605834 MIM: 600607 | ||||||||||||||||||||
Literature Links: |
TMOD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMOD4 - tropomodulin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS72 - vacuolar protein sorting 72 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271087.1 | 423 | Missense Mutation | ACG,GCG | T,A 111 | NP_001258016.1 | |
NM_001271088.1 | 423 | Missense Mutation | ACG,GCG | T,A 111 | NP_001258017.1 | |
NM_005997.2 | 423 | Missense Mutation | ACG,GCG | T,A 111 | NP_005988.1 | |
XM_017002205.1 | 423 | UTR 5 | XP_016857694.1 |