Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGCCCCTGGCCACGTGGTCACT[C/T]ATGAAGGTGGTCCTTCCTCTGTACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
BTNL10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BTNL10 - butyrophilin like 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287262.1 | 327 | Missense Mutation | ATA,ATG | I,M 58 | NP_001274191.1 | |
XM_011544015.2 | 327 | Missense Mutation | ATA,ATG | I,M 58 | XP_011542317.1 | |
XM_016999986.1 | 327 | Missense Mutation | ATA,ATG | I,M 58 | XP_016855475.1 | |
XM_016999987.1 | 327 | Missense Mutation | ATA,ATG | I,M 58 | XP_016855476.1 |