Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGTGCCCAGACCACAAGGAAGAG[A/G]TGACCCACTACTGCAAGACATGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 158340 MIM: 600986 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRTCAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRTCAP2 - keratinocyte associated protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MUC1 - mucin 1, cell surface associated | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM46 - tripartite motif containing 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256599.1 | 994 | Missense Mutation | ATG,GTG | M,V 211 | NP_001243528.1 | |
NM_001256600.1 | 994 | Missense Mutation | ATG,GTG | M,V 221 | NP_001243529.1 | |
NM_001256601.1 | 994 | Missense Mutation | ATG,GTG | M,V 221 | NP_001243530.1 | |
NM_001282378.1 | 994 | Missense Mutation | ATG,GTG | M,V 108 | NP_001269307.1 | |
NM_001282379.1 | 994 | Missense Mutation | ATG,GTG | M,V 234 | NP_001269308.1 | |
NM_025058.4 | 994 | Missense Mutation | ATG,GTG | M,V 234 | NP_079334.3 | |
XM_011510002.1 | 994 | Missense Mutation | ATG,GTG | M,V 263 | XP_011508304.1 | |
XM_017002393.1 | 994 | Missense Mutation | ATG,GTG | M,V 221 | XP_016857882.1 |