Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATTGCTGTGGACCGCTACCTCCG[G/T]GTCAAGATCCCTCTCCGGTGAGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 102775 | ||||||||||||||||||||
Literature Links: |
ADORA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADORA1 - adenosine A1 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000674.2 | 252 | Silent Mutation | CGG,CGT | R,R 108 | NP_000665.1 | |
NM_001048230.1 | 252 | Silent Mutation | CGG,CGT | R,R 108 | NP_001041695.1 | |
XM_005244901.1 | 252 | UTR 5 | XP_005244958.1 | |||
XM_005244902.4 | 252 | UTR 5 | XP_005244959.1 | |||
XM_017000342.1 | 252 | UTR 5 | XP_016855831.1 |