Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGAGAGCCATGTCGGCCAGCAGC[C/G]TCTTGGAGCAGGTACAGGCCCGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610640 | ||||||||||||||||||||
Literature Links: |
YTHDF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
YTHDF2 - YTH N6-methyladenosine RNA binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172828.1 | 513 | UTR 5 | NP_001166299.1 | |||
NM_001173128.1 | 513 | Missense Mutation | CTC,GTC | L,V 6 | NP_001166599.1 | |
NM_016258.2 | 513 | Missense Mutation | CTC,GTC | L,V 6 | NP_057342.2 |