Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGAACCTCTGGCGCCCTGCTG[C/T]TCCCAGGGGCGTCTAGCACCGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616129 MIM: 606822 MIM: 613170 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LURAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LURAP1 - leucine rich adaptor protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013615.2 | 226 | Missense Mutation | CTC,TTC | L,F 45 | NP_001013633.1 |
POMGNT1 - protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243766.1 | 226 | Intron | NP_001230695.1 | |||
NM_001290129.1 | 226 | Intron | NP_001277058.1 | |||
NM_001290130.1 | 226 | Intron | NP_001277059.1 | |||
NM_017739.3 | 226 | Intron | NP_060209.3 | |||
XM_005271010.1 | 226 | Intron | XP_005271067.1 | |||
XM_006710755.1 | 226 | Intron | XP_006710818.1 | |||
XM_006710756.1 | 226 | Intron | XP_006710819.1 | |||
XM_011541760.2 | 226 | Intron | XP_011540062.1 | |||
XM_017001690.1 | 226 | Intron | XP_016857179.1 |
TSPAN1 - tetraspanin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |