Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTCTTACAGTGGTACTCGTCAC[A/G]AGCTTCTCGGTGGACAAGCAACATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603341 MIM: 600024 | ||||||||||||||||||||
Literature Links: |
DNAH14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAH14 - dynein axonemal heavy chain 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LBR - lamin B receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002296.3 | 1760 | Missense Mutation | CGT,TGT | R,C 586 | NP_002287.2 | |
NM_194442.2 | 1760 | Missense Mutation | CGT,TGT | R,C 586 | NP_919424.1 | |
XM_005273125.2 | 1760 | Missense Mutation | CGT,TGT | R,C 544 | XP_005273182.1 | |
XM_011544185.2 | 1760 | Missense Mutation | CGT,TGT | R,C 586 | XP_011542487.1 |