Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTAGGTTTACTGACATTCTTCAA[C/T]CGTGGATTTGAATTTTGTTGTTTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601848 | ||||||||||||||||||||
Literature Links: |
RBM34 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RBM34 - RNA binding motif protein 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161533.1 | 1458 | Intron | NP_001155005.1 | |||
NM_015014.2 | 1458 | Silent Mutation | CGA,CGG | R,R 376 | NP_055829.2 | |
XM_011544133.1 | 1458 | Silent Mutation | CGA,CGG | R,R 356 | XP_011542435.1 | |
XM_017000721.1 | 1458 | Silent Mutation | CGA,CGG | R,R 375 | XP_016856210.1 |
SNORA14B - small nucleolar RNA, H/ACA box 14B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOMM20 - translocase of outer mitochondrial membrane 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |