Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCTTTCCCACACTGCACACACTC[A/G]TAGGGCTTCTCCCCTGTGTGGGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611703 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF436 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF436 - zinc finger protein 436 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077195.1 | 1813 | Silent Mutation | TAC,TAT | Y,Y 418 | NP_001070663.1 | |
NM_030634.2 | 1813 | Silent Mutation | TAC,TAT | Y,Y 418 | NP_085137.1 | |
XM_011542201.1 | 1813 | Silent Mutation | TAC,TAT | Y,Y 400 | XP_011540503.1 |
ZNF436-AS1 - ZNF436 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |