Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCTCTCACCTCCGTTCCTCGGTC[A/G]ATGGCTACTTTGCCCAGCCGCACGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ECHDC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ECHDC2 - enoyl-CoA hydratase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198961.1 | 4734 | Silent Mutation | ATC,ATT | I,I 246 | NP_001185890.1 | |
NM_001198962.1 | 4734 | Intron | NP_001185891.1 | |||
NM_001319958.1 | 4734 | Silent Mutation | ATC,ATT | I,I 169 | NP_001306887.1 | |
NM_018281.3 | 4734 | Silent Mutation | ATC,ATT | I,I 215 | NP_060751.2 | |
XM_011541709.2 | 4734 | Silent Mutation | ATC,ATT | I,I 246 | XP_011540011.1 | |
XM_011541710.2 | 4734 | Silent Mutation | ATC,ATT | I,I 246 | XP_011540012.1 | |
XM_011541711.2 | 4734 | Silent Mutation | ATC,ATT | I,I 215 | XP_011540013.1 | |
XM_011541712.2 | 4734 | Silent Mutation | ATC,ATT | I,I 246 | XP_011540014.1 | |
XM_011541713.2 | 4734 | Silent Mutation | ATC,ATT | I,I 200 | XP_011540015.1 | |
XM_011541715.2 | 4734 | Silent Mutation | ATC,ATT | I,I 200 | XP_011540017.1 | |
XM_011541719.2 | 4734 | Silent Mutation | ATC,ATT | I,I 200 | XP_011540021.1 | |
XM_011541720.2 | 4734 | Silent Mutation | ATC,ATT | I,I 200 | XP_011540022.1 | |
XM_011541721.2 | 4734 | Silent Mutation | ATC,ATT | I,I 200 | XP_011540023.1 | |
XM_011541722.2 | 4734 | Silent Mutation | ATC,ATT | I,I 200 | XP_011540024.1 | |
XM_011541723.2 | 4734 | Silent Mutation | ATC,ATT | I,I 200 | XP_011540025.1 | |
XM_011541724.2 | 4734 | Silent Mutation | ATC,ATT | I,I 196 | XP_011540026.1 | |
XM_011541725.2 | 4734 | Silent Mutation | ATC,ATT | I,I 215 | XP_011540027.1 | |
XM_011541726.2 | 4734 | Silent Mutation | ATC,ATT | I,I 169 | XP_011540028.1 | |
XM_011541727.2 | 4734 | Silent Mutation | ATC,ATT | I,I 150 | XP_011540029.1 | |
XM_017001638.1 | 4734 | Silent Mutation | ATC,ATT | I,I 200 | XP_016857127.1 | |
XM_017001639.1 | 4734 | Silent Mutation | ATC,ATT | I,I 200 | XP_016857128.1 | |
XM_017001640.1 | 4734 | Silent Mutation | ATC,ATT | I,I 200 | XP_016857129.1 | |
XM_017001641.1 | 4734 | Silent Mutation | ATC,ATT | I,I 200 | XP_016857130.1 | |
XM_017001642.1 | 4734 | Silent Mutation | ATC,ATT | I,I 169 | XP_016857131.1 |
ZYG11A - zyg-11 family member A, cell cycle regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |