Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAAACTTTTACAAAGAAGAATTC[C/T]TTTTCTGTAGCCACTGTAGCATGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604578 | ||||||||||||||||||||
Literature Links: |
AIM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AIM2 - absent in melanoma 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004833.1 | 800 | Silent Mutation | AAA,AAG | K,K 185 | NP_004824.1 | |
XM_005245616.4 | 800 | Silent Mutation | AAA,AAG | K,K 80 | XP_005245673.1 | |
XM_017002848.1 | 800 | Silent Mutation | AAA,AAG | K,K 185 | XP_016858337.1 |