Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTACGGGACCGGGACCGGTACCG[C/T]GAAGGAGACCGGTAGTATCTCCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RSRP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RSRP1 - arginine and serine rich protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321772.1 | 569 | Silent Mutation | TCA,TCG | S,S 109 | NP_001308701.1 | |
NM_020317.4 | 569 | Silent Mutation | TCA,TCG | S,S 109 | NP_064713.3 | |
XM_011541797.1 | 569 | Silent Mutation | TCA,TCG | S,S 109 | XP_011540099.1 |