Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGCCTACAGGCTGGACAGATGG[C/T]GGGGGGCCCCTGGGGTGAGCGACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602344 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAFAH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAFAH2 - platelet activating factor acetylhydrolase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000437.3 | 1093 | Missense Mutation | CAC,CGC | H,R 387 | NP_000428.2 | |
XM_006710669.3 | 1093 | Missense Mutation | CAC,CGC | H,R 280 | XP_006710732.1 | |
XM_006710670.3 | 1093 | Missense Mutation | CAC,CGC | H,R 387 | XP_006710733.1 | |
XM_011541528.2 | 1093 | Missense Mutation | CAC,CGC | H,R 411 | XP_011539830.1 | |
XM_011541529.2 | 1093 | Missense Mutation | CAC,CGC | H,R 266 | XP_011539831.1 | |
XM_017001390.1 | 1093 | Missense Mutation | CAC,CGC | H,R 266 | XP_016856879.1 | |
XM_017001391.1 | 1093 | Missense Mutation | CAC,CGC | H,R 266 | XP_016856880.1 | |
XM_017001392.1 | 1093 | Missense Mutation | CAC,CGC | H,R 266 | XP_016856881.1 |