Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGTTTGTTTTTCTGCATACTTAA[C/T]TCCAATGTCAATGGTTGTATGGAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602374 | ||||||||||||||||||||
Literature Links: |
CNN3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNN3 - calponin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286055.1 | 543 | Missense Mutation | ATT,GTT | I,V 96 | NP_001272984.1 | |
NM_001286056.1 | 543 | Missense Mutation | ATT,GTT | I,V 101 | NP_001272985.1 | |
NM_001839.4 | 543 | Missense Mutation | ATT,GTT | I,V 142 | NP_001830.1 | |
XM_017000245.1 | 543 | Missense Mutation | ATT,GTT | I,V 101 | XP_016855734.1 |
SLC44A3 - solute carrier family 44 member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |