Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTGGCATTCCCTGGGGACTTGT[C/T]TGGGCATAATAGGCTGCTTGTTGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603444 MIM: 613121 | ||||||||||||||||||||
Literature Links: |
FUBP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FUBP1 - far upstream element binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303433.1 | 2050 | Silent Mutation | CAA,CAG | Q,Q 649 | NP_001290362.1 | |
NM_003902.4 | 2050 | Silent Mutation | CAA,CAG | Q,Q 628 | NP_003893.2 | |
XM_011542391.2 | 2050 | Silent Mutation | CAA,CAG | Q,Q 649 | XP_011540693.1 | |
XM_011542392.2 | 2050 | Silent Mutation | CAA,CAG | Q,Q 648 | XP_011540694.1 | |
XM_011542393.2 | 2050 | Silent Mutation | CAA,CAG | Q,Q 628 | XP_011540695.1 | |
XM_017002739.1 | 2050 | Silent Mutation | CAA,CAG | Q,Q 649 | XP_016858228.1 | |
XM_017002740.1 | 2050 | Silent Mutation | CAA,CAG | Q,Q 649 | XP_016858229.1 | |
XM_017002741.1 | 2050 | Silent Mutation | CAA,CAG | Q,Q 627 | XP_016858230.1 | |
XM_017002742.1 | 2050 | Silent Mutation | CAA,CAG | Q,Q 648 | XP_016858231.1 | |
XM_017002743.1 | 2050 | Silent Mutation | CAA,CAG | Q,Q 627 | XP_016858232.1 | |
XM_017002744.1 | 2050 | Silent Mutation | CAA,CAG | Q,Q 627 | XP_016858233.1 |
NEXN - nexilin F-actin binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172309.1 | 2050 | Intron | NP_001165780.1 | |||
NM_144573.3 | 2050 | Intron | NP_653174.3 |