Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGGACCCACAGGTGGAGCGGGC[A/G]CTGGCGGCCGTAGAGCGTCAGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 121012 MIM: 603324 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GJA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GJA4 - gap junction protein alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002060.2 | 635 | Silent Mutation | GCA,GCG | A,A 126 | NP_002051.2 | |
XM_005270750.2 | 635 | Silent Mutation | GCA,GCG | A,A 126 | XP_005270807.1 | |
XM_017001043.1 | 635 | Silent Mutation | GCA,GCG | A,A 126 | XP_016856532.1 |
GJB3 - gap junction protein beta 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |