Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTCCACAGGAGTACACCATGAC[A/G]GTGTTCCTGCACCAGAGCTGGCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137163 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABRD - gamma-aminobutyric acid type A receptor delta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000815.4 | 359 | Silent Mutation | ACA,ACG | T,T 88 | NP_000806.2 | |
XM_011541194.2 | 359 | Silent Mutation | ACA,ACG | T,T 101 | XP_011539496.1 | |
XM_017000936.1 | 359 | Silent Mutation | ACA,ACG | T,T 323 | XP_016856425.1 |
LOC105378589 - uncharacterized LOC105378589 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |