Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCCTGTGCTCTTTGTCTTTCTTC[C/T]TCTTCTCTTTCTTGTGTTTCCTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 103180 MIM: 611859 | ||||||||||||||||||||
Literature Links: |
ARF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARF1 - ADP ribosylation factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C1orf35 - chromosome 1 open reading frame 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024319.3 | 840 | Missense Mutation | AAG,AGG | K,R 202 | NP_077295.1 |
MIR3620 - microRNA 3620 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL55 - mitochondrial ribosomal protein L55 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |