Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTATTTCAGGACTGGAAAGGAA[G/T]GCCAACCCAGAGAGTACTACACATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607393 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC73 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDC73 - cell division cycle 73 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024529.4 | 388 | Missense Mutation | GGC,TGC | G,C 49 | NP_078805.3 | |
XM_006711537.3 | 388 | Missense Mutation | GGC,TGC | G,C 49 | XP_006711600.1 |