Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCTGTAGGACCAGGGGGTGTTGG[C/G]GAGGAAGACTGCAGGAGGGGAGCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609034 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEYL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HEYL - hes related family bHLH transcription factor with YRPW motif-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014571.3 | 891 | Silent Mutation | TCC,TCG | S,S 280 | NP_055386.1 | |
XM_005270745.2 | 891 | Silent Mutation | TCC,TCG | S,S 252 | XP_005270802.1 |