Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCTGGTAGCTCAGGCGGCCGAT[A/G]TACACCCGCGGCATCCCGGCAACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601940 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRSF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SRSF4 - serine and arginine rich splicing factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005626.4 | 188 | Silent Mutation | TAC,TAT | Y,Y 5 | NP_005617.2 | |
XM_011541951.2 | 188 | Silent Mutation | TAC,TAT | Y,Y 5 | XP_011540253.1 |