Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAATTGCAAGCCCAGGCATCTC[C/T]GCAGCCAGAGCCTGAAGAACTCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611315 | ||||||||||||||||||||
Literature Links: |
ZSCAN20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZSCAN20 - zinc finger and SCAN domain containing 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145238.3 | 191 | Missense Mutation | CCG,CTG | P,L 13 | NP_660281.2 | |
XM_005271171.3 | 191 | Missense Mutation | CCG,CTG | P,L 13 | XP_005271228.1 | |
XM_006710874.3 | 191 | Missense Mutation | CCG,CTG | P,L 13 | XP_006710937.1 | |
XM_006710876.3 | 191 | Missense Mutation | CCG,CTG | P,L 13 | XP_006710939.1 | |
XM_017002237.1 | 191 | Missense Mutation | CCG,CTG | P,L 13 | XP_016857726.1 | |
XM_017002238.1 | 191 | Missense Mutation | CCG,CTG | P,L 13 | XP_016857727.1 | |
XM_017002239.1 | 191 | UTR 5 | XP_016857728.1 | |||
XM_017002240.1 | 191 | UTR 5 | XP_016857729.1 | |||
XM_017002241.1 | 191 | Intron | XP_016857730.1 |