Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTAGACAAGTCCACACCCTACA[C/T]GGCTGTGCGATGGGTCGTGACACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602859 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MORN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MORN1 - MORN repeat containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PEX10 - peroxisomal biogenesis factor 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RER1 - retention in endoplasmic reticulum sorting receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007033.4 | 417 | Missense Mutation | ACG,ATG | T,M 40 | NP_008964.3 | |
XM_005244713.4 | 417 | Missense Mutation | ACG,ATG | T,M 68 | XP_005244770.2 | |
XM_006710306.2 | 417 | Missense Mutation | ACG,ATG | T,M 40 | XP_006710369.1 | |
XM_011540542.1 | 417 | Missense Mutation | ACG,ATG | T,M 40 | XP_011538844.1 | |
XM_011540543.1 | 417 | Missense Mutation | ACG,ATG | T,M 40 | XP_011538845.1 | |
XM_017000131.1 | 417 | Missense Mutation | ACG,ATG | T,M 75 | XP_016855620.1 | |
XM_017000132.1 | 417 | Missense Mutation | ACG,ATG | T,M 75 | XP_016855621.1 | |
XM_017000133.1 | 417 | Missense Mutation | ACG,ATG | T,M 40 | XP_016855622.1 | |
XM_017000134.1 | 417 | Missense Mutation | ACG,ATG | T,M 40 | XP_016855623.1 | |
XM_017000135.1 | 417 | Missense Mutation | ACG,ATG | T,M 40 | XP_016855624.1 |