Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGAGAAAGGCGAATACATTCAC[G/T]TCACCCCTGACCTTCTACTTGGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616857 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLCA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLCA4 - chloride channel accessory 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012128.3 | 441 | Missense Mutation | GTC,TTC | V,F 133 | NP_036260.2 | |
XM_011541015.2 | 441 | Missense Mutation | GTC,TTC | V,F 82 | XP_011539317.1 |