Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACCTGGGCTCAGTGGCAGATCC[G/T]CTCTGTCATCTCCCTCTGTAGAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606724 | ||||||||||||||||||||
Literature Links: |
MKNK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MKNK1 - MAP kinase interacting serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MOB3C - MOB kinase activator 3C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145279.4 | 3799 | Silent Mutation | AGG,CGG | R,R 265 | NP_660322.2 | |
NM_201403.2 | 3799 | Silent Mutation | AGG,CGG | R,R 213 | NP_958805.1 | |
XM_017000407.1 | 3799 | Silent Mutation | AGG,CGG | R,R 236 | XP_016855896.1 |